Product Details

SNP ID
rs13230797
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143913141 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGATCGAGATAATATATTCAAGC[A/C]CATGAAAGAGACAAAGAAATATCAT
Phenotype
MIM: 608497
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR2F1 PubMed Links

Gene Details

Gene
OR2F1
Gene Name
olfactory receptor family 2 subfamily F member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012369.2 Intron NP_036501.2
XM_011516019.2 Intron XP_011514321.1
XM_011516020.2 Intron XP_011514322.1

View Full Product Details