Product Details
- SNP ID
-
rs13343388
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:42430660 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCAACAGCACCCCTCCTTTTCTG[A/T]TCACTGCTTTCTCTCCCCCTCACAG
- Phenotype
-
MIM: 611387
MIM: 151750
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CXCL17
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs537870150,rs538300279] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CXCL17
- Gene Name
- C-X-C motif chemokine ligand 17
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_198477.2 |
|
Intron |
|
|
NP_940879.1 |
- Gene
- LIPE
- Gene Name
- lipase E, hormone sensitive type
There are no transcripts associated with this gene.
- Gene
- LIPE-AS1
- Gene Name
- LIPE antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details