Product Details
- SNP ID
-
rs13308178
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:44073666 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCCCATGGCTGTTCAGCCACAGGC[C/G]CTTCTCCTTCCGGCTGAAGCGGCGC
- Phenotype
-
MIM: 612931
MIM: 606344
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIR6838
PubMed Links
Gene Details
- Gene
- MIR6838
- Gene Name
- microRNA 6838
There are no transcripts associated with this gene.
- Gene
- PGAM2
- Gene Name
- phosphoglycerate mutase 2
There are no transcripts associated with this gene.
- Gene
- POLM
- Gene Name
- DNA polymerase mu
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001284330.1 |
1314 |
Missense Mutation |
CGC,GGC |
R373G |
NP_001271259.1 |
NM_001284331.1 |
1314 |
Missense Mutation |
CGC,GGC |
R416G |
NP_001271260.1 |
NM_013284.3 |
1314 |
Missense Mutation |
CGC,GGC |
R453G |
NP_037416.1 |
XM_005249711.2 |
1314 |
Missense Mutation |
CGC,GGC |
R313G |
XP_005249768.1 |
XM_006715692.2 |
1314 |
Missense Mutation |
CGC,GGC |
R453G |
XP_006715755.1 |
XM_006715696.2 |
1314 |
Missense Mutation |
CGC,GGC |
R363G |
XP_006715759.1 |
XM_006715698.3 |
1314 |
Missense Mutation |
CGC,GGC |
R226G |
XP_006715761.1 |
XM_011515275.1 |
1314 |
Missense Mutation |
CGC,GGC |
R458G |
XP_011513577.1 |
XM_011515278.1 |
1314 |
Missense Mutation |
CGC,GGC |
R408G |
XP_011513580.1 |
XM_011515279.1 |
1314 |
Missense Mutation |
CGC,GGC |
R458G |
XP_011513581.1 |
XM_011515282.2 |
1314 |
Missense Mutation |
CGC,GGC |
R225G |
XP_011513584.1 |
XM_011515285.1 |
1314 |
Missense Mutation |
CGC,GGC |
R189G |
XP_011513587.1 |
XM_011515286.2 |
1314 |
Intron |
|
|
XP_011513588.1 |
XM_011515287.2 |
1314 |
Intron |
|
|
XP_011513589.1 |
XM_017011998.1 |
1314 |
Missense Mutation |
CGC,GGC |
R396G |
XP_016867487.1 |
XM_017011999.1 |
1314 |
Missense Mutation |
CGC,GGC |
R378G |
XP_016867488.1 |
XM_017012000.1 |
1314 |
Missense Mutation |
CGC,GGC |
R408G |
XP_016867489.1 |
XM_017012001.1 |
1314 |
Missense Mutation |
CGC,GGC |
R378G |
XP_016867490.1 |
XM_017012002.1 |
1314 |
Missense Mutation |
CGC,GGC |
R189G |
XP_016867491.1 |
XM_017012003.1 |
1314 |
Missense Mutation |
CGC,GGC |
R189G |
XP_016867492.1 |
XM_017012004.1 |
1314 |
Intron |
|
|
XP_016867493.1 |
XM_017012005.1 |
1314 |
Intron |
|
|
XP_016867494.1 |
View Full Product Details