Product Details

SNP ID
rs13174563
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:139276006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACAAGGATAAGCTATTGGTTTTA[A/T]TGTCTGCTATTGTTTCTGTGGCTTA
Phenotype
MIM: 164015
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MATR3 PubMed Links

Gene Details

Gene
MATR3
Gene Name
matrin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001194954.1 Intron NP_001181883.1
NM_001194955.1 Intron NP_001181884.1
NM_001194956.1 Intron NP_001181885.1
NM_001282278.1 Intron NP_001269207.1
NM_018834.5 Intron NP_061322.2
NM_199189.2 Intron NP_954659.1
Gene
SNHG4
Gene Name
small nucleolar RNA host gene 4
There are no transcripts associated with this gene.

Gene
SNORA74A
Gene Name
small nucleolar RNA, H/ACA box 74A
There are no transcripts associated with this gene.

View Full Product Details