Product Details
- SNP ID
-
rs10958572
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:39937854 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTATTTTAATGAGACAACAAAGAT[A/C]AAGGGAACTTTAGTCCTTACAATGT
- Phenotype
-
MIM: 147435
MIM: 612129
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
IDO1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116691269] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IDO1
- Gene Name
- indoleamine 2,3-dioxygenase 1
There are no transcripts associated with this gene.
- Gene
- IDO2
- Gene Name
- indoleamine 2,3-dioxygenase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_194294.2 |
|
Intron |
|
|
NP_919270.2 |
View Full Product Details