Product Details

SNP ID
rs11708257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:47016567 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAAACCACAAGGCAGCCTGCTTT[A/G]GAATATTTACATGATAACAAATTAA
Phenotype
MIM: 614169 MIM: 612778
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NBEAL2 PubMed Links

Gene Details

Gene
NBEAL2
Gene Name
neurobeachin like 2
There are no transcripts associated with this gene.

Gene
NRADDP
Gene Name
neurotrophin receptor associated death domain, pseudogene
There are no transcripts associated with this gene.

Gene
SETD2
Gene Name
SET domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014159.6 9770 UTR 3 NP_054878.5
XM_011533631.2 9770 UTR 3 XP_011531933.1
XM_011533632.2 9770 UTR 3 XP_011531934.1
XM_011533633.2 9770 UTR 3 XP_011531935.1
XM_011533634.1 9770 UTR 3 XP_011531936.1
XM_017006270.1 9770 UTR 3 XP_016861759.1

View Full Product Details