Product Details

SNP ID
rs11038984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:46745201 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGAATGAACAATGATACAATGC[A/G]GGCCTTTAAAATATTTCACTTGGTA
Phenotype
MIM: 611142 MIM: 176930
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CKAP5 PubMed Links

Gene Details

Gene
CKAP5
Gene Name
cytoskeleton associated protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008938.3 Intron NP_001008938.1
NM_014756.3 Intron NP_055571.2
Gene
F2
Gene Name
coagulation factor II, thrombin
There are no transcripts associated with this gene.

Gene
MIR5582
Gene Name
microRNA 5582
There are no transcripts associated with this gene.

View Full Product Details