Product Details

SNP ID
rs12283996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6681654 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACTTGCAGATTGTCACTCCCCTA[C/G]CCCTGTCAGGAAGTAATCTTTCTCT
Phenotype
MIM: 611830
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MRPL17 PubMed Links

Gene Details

Gene
MRPL17
Gene Name
mitochondrial ribosomal protein L17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022061.3 1097 UTR 3 NP_071344.1

View Full Product Details