Product Details

SNP ID
rs10897341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63080895 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGCACATAGCCACTCCCCACTCA[A/C]GTGACAGCTAGAGGGTTTACTCACT
Phenotype
MIM: 611698
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC22A24 PubMed Links

Gene Details

Gene
SLC22A24
Gene Name
solute carrier family 22 member 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136506.2 Intron NP_001129978.2
NM_173586.2 Intron NP_775857.2
XM_011544967.2 Intron XP_011543269.1

View Full Product Details