Product Details

SNP ID
rs13240993
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:144397318 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGATCCAGGGAAGCAGCTGGTGG[G/T]TCCTCTTTTGCCTTGCTGAGTAAGG
Phenotype
MIM: 610934
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NOBOX PubMed Links

Gene Details

Gene
NOBOX
Gene Name
NOBOX oogenesis homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080413.3 2113 Missense Mutation GAA,GAC E666D NP_001073882.3
XM_017011742.1 2113 Missense Mutation GAA,GAC E634D XP_016867231.1

View Full Product Details