Product Details

SNP ID
rs11644212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:50153889 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGCATGTATCGCTCCGGGGAGCGC[A/C]TGCTGGGCAGCCACGCGCTGCCCGC
Phenotype
MIM: 605540
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PAPD5 PubMed Links

Gene Details

Gene
PAPD5
Gene Name
PAP associated domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040284.2 314 Missense Mutation ATG,CTG M75L NP_001035374.2
NM_001040285.2 314 Missense Mutation ATG,CTG M75L NP_001035375.2
XM_005256092.4 314 Missense Mutation ATG,CTG M90L XP_005256149.1
XM_005256093.4 314 Missense Mutation ATG,CTG M76L XP_005256150.2
XM_011523275.2 314 Missense Mutation ATG,CTG M76L XP_011521577.1
XM_011523276.2 314 Missense Mutation ATG,CTG M76L XP_011521578.1
XM_011523278.2 314 Intron XP_011521580.1
XM_017023555.1 314 Missense Mutation ATG,CTG M70L XP_016879044.1

View Full Product Details