Product Details

SNP ID
rs12354141
Assay Type
Functionally tested
NCBI dbSNP Submissions
40
Location
Chr.1:174166078 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTAACATTGAACAGGATTTATAT[A/G]TTATGATTAAAATATGATCCAGAGA
Phenotype
MIM: 609238
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC102724601 PubMed Links

Gene Details

Gene
LOC102724601
Gene Name
uncharacterized LOC102724601
There are no transcripts associated with this gene.

Gene
RABGAP1L
Gene Name
RAB GTPase activating protein 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035230.2 Intron NP_001030307.1
NM_001243763.1 Intron NP_001230692.1
NM_001243764.1 Intron NP_001230693.1
NM_001243765.1 Intron NP_001230694.1
NM_014857.4 Intron NP_055672.3
XM_005245680.1 Intron XP_005245737.1
XM_005245681.2 Intron XP_005245738.1
XM_006711693.1 Intron XP_006711756.1
XM_011510223.2 Intron XP_011508525.1
XM_017002994.1 Intron XP_016858483.1
XM_017002995.1 Intron XP_016858484.1
XM_017002996.1 Intron XP_016858485.1
XM_017002997.1 Intron XP_016858486.1
XM_017002998.1 Intron XP_016858487.1
XM_017002999.1 Intron XP_016858488.1

View Full Product Details