Product Details

SNP ID
rs10912737
Assay Type
Functionally tested
NCBI dbSNP Submissions
39
Location
Chr.1:174172999 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTAAGGTTTTATTATAGATATTA[A/G]TGTTTACTTCTTTAAGCTATTTTAG
Phenotype
MIM: 609238
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RABGAP1L PubMed Links

Gene Details

Gene
RABGAP1L
Gene Name
RAB GTPase activating protein 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035230.2 Intron NP_001030307.1
NM_001243763.1 Intron NP_001230692.1
NM_001243764.1 Intron NP_001230693.1
NM_001243765.1 Intron NP_001230694.1
NM_014857.4 Intron NP_055672.3
XM_005245680.1 Intron XP_005245737.1
XM_005245681.2 Intron XP_005245738.1
XM_006711693.1 Intron XP_006711756.1
XM_011510223.2 Intron XP_011508525.1
XM_017002994.1 Intron XP_016858483.1
XM_017002995.1 Intron XP_016858484.1
XM_017002996.1 Intron XP_016858485.1
XM_017002997.1 Intron XP_016858486.1
XM_017002998.1 Intron XP_016858487.1
XM_017002999.1 Intron XP_016858488.1

View Full Product Details