Product Details

SNP ID
rs12087100
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:201987189 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGCCTCTCCTCATGTTGCTGCTC[A/G]TCTCCTTGTGAATGTCTCCTCCCTT
Phenotype
MIM: 602675
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNPEP PubMed Links

Gene Details

Gene
RNPEP
Gene Name
arginyl aminopeptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319182.1 Intron NP_001306111.1
NM_001319183.1 Intron NP_001306112.1
NM_001319184.1 Intron NP_001306113.1
NM_020216.3 Intron NP_064601.3
XM_005245421.1 Intron XP_005245478.1
XM_017002022.1 Intron XP_016857511.1

View Full Product Details