Product Details

SNP ID
rs6980482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:109968831 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTGAGAAGGCTAGAGAGAAATATA[A/G]TTAGATTCATTAATTCACCTATAGA
Phenotype
MIM: 608164
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNV1 PubMed Links

Gene Details

Gene
KCNV1
Gene Name
potassium voltage-gated channel modifier subfamily V member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014379.3 Intron NP_055194.1

View Full Product Details