Product Details

SNP ID
rs13300924
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:96390378 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGATGACTAGAAGAGGAGAATGG[C/T]AAAAGATAAGTTCAGCAAGTCAGCA
Phenotype
MIM: 609182 MIM: 610160
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC35D2 PubMed Links

Gene Details

Gene
SLC35D2
Gene Name
solute carrier family 35 member D2
There are no transcripts associated with this gene.

Gene
ZNF367
Gene Name
zinc finger protein 367
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153695.3 Intron NP_710162.1
XM_011518345.2 Intron XP_011516647.1

View Full Product Details