Product Details

SNP ID
rs13163338
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:77077519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCGCCCTCGGGGGCCGCAGCGGG[G/T]CCGGGCGGCGGCGGGCAGGGCGCGG
Phenotype
MIM: 615250
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SNORA47 PubMed Links

Gene Details

Gene
SNORA47
Gene Name
small nucleolar RNA, H/ACA box 47
There are no transcripts associated with this gene.

Gene
ZBED3
Gene Name
zinc finger BED-type containing 3
There are no transcripts associated with this gene.

Gene
ZBED3-AS1
Gene Name
ZBED3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details