Product Details

SNP ID
rs11068213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:116860511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGAGGAGGCTGTGGGCTGAACT[C/T]CGAGGACACTTTTGTGGGGGTGGGG
Phenotype
MIM: 603447
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HRK PubMed Links

Gene Details

Gene
HRK
Gene Name
harakiri, BCL2 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003806.2 1716 UTR 3 NP_003797.1
XM_017020146.1 1716 UTR 3 XP_016875635.1
Gene
RNFT2
Gene Name
ring finger protein, transmembrane 2
There are no transcripts associated with this gene.

View Full Product Details