Product Details

SNP ID
rs11068214
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:116862556 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGAGAATGGTGGTTGCCTAGGGC[C/T]GGGGCGGATAGGAGGAATTGGGGAG
Phenotype
MIM: 603447
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HRK PubMed Links

Gene Details

Gene
HRK
Gene Name
harakiri, BCL2 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003806.2 Intron NP_003797.1
XM_017020146.1 Intron XP_016875635.1
Gene
RNFT2
Gene Name
ring finger protein, transmembrane 2
There are no transcripts associated with this gene.

View Full Product Details