Product Details

SNP ID
rs9658574
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:117212919 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCAGATCGCTCTCCTGCCTTCTAG[C/T]CTGGAGTTGTGAAGCAGCTTGTGTT
Phenotype
MIM: 163731
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NOS1 PubMed Links

Gene Details

Gene
NOS1
Gene Name
nitric oxide synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000620.4 7515 UTR 3 NP_000611.1
NM_001204213.1 7515 UTR 3 NP_001191142.1
NM_001204214.1 7515 UTR 3 NP_001191143.1
NM_001204218.1 7515 UTR 3 NP_001191147.1
XM_011538398.2 7515 UTR 3 XP_011536700.1
XM_017019345.1 7515 UTR 3 XP_016874834.1
XM_017019346.1 7515 UTR 3 XP_016874835.1
XM_017019347.1 7515 UTR 3 XP_016874836.1

View Full Product Details