Product Details
- SNP ID
-
hCV31136398
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:122380155 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAAGAACCTGCCAGTTCAACCAG[C/G]GACAAAGACGATAACTACACCCACA
- Phenotype
-
MIM: 614130
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ADAD1
PubMed Links
Gene Details
- Gene
- ADAD1
- Gene Name
- adenosine deaminase domain containing 1
There are no transcripts associated with this gene.
View Full Product Details