Product Details
- SNP ID
-
rs12510548
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:121669280 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAAATAAAAGAAGGACTAATAAAGA[C/G]AGCTGGAATTGATAAAAGACAGAAA
- Phenotype
-
MIM: 131230
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ANXA5
PubMed Links
Gene Details
- Gene
- ANXA5
- Gene Name
- annexin A5
View Full Product Details