Product Details

SNP ID
rs6929766
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:158176201 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAACGTTACATATACTATCTCTCC[A/C]TTATAAAACTAAGTATAATATTGTG
Phenotype
MIM: 608780 MIM: 614725
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GTF2H5 PubMed Links

Gene Details

Gene
GTF2H5
Gene Name
general transcription factor IIH subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207118.2 Intron NP_997001.1
XM_017010862.1 Intron XP_016866351.1
Gene
SERAC1
Gene Name
serine active site containing 1
There are no transcripts associated with this gene.

View Full Product Details