Product Details

SNP ID
rs4687396
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:193241984 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAGTTACTTGTGCGAGGCACTCT[A/C]ACCGGAGACTGAAGGTATAGAGCAA
Phenotype
MIM: 606487
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
HRASLS PubMed Links

Gene Details

Gene
HRASLS
Gene Name
HRAS like suppressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020386.4 Intron NP_065119.2
XM_011513034.1 Intron XP_011511336.1
XM_011513035.2 Intron XP_011511337.1
XM_011513037.2 Intron XP_011511339.1
XM_017006923.1 Intron XP_016862412.1
Gene
MGC2889
Gene Name
uncharacterized protein MGC2889
There are no transcripts associated with this gene.

View Full Product Details