Product Details

SNP ID
rs10901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:86206117 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTATCTTTACAGGAATCCTACGG[C/T]CCCTGAATATTTTGGCATCTTCAAC
Phenotype
MIM: 611841 MIM: 609139
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL35 PubMed Links

Gene Details

Gene
MRPL35
Gene Name
mitochondrial ribosomal protein L35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016622.3 113 Missense Mutation CCC,TCC P19S NP_057706.2
NM_145644.2 113 Missense Mutation CCC,TCC P19S NP_663619.1
XM_005264361.2 113 Intron XP_005264418.1
Gene
REEP1
Gene Name
receptor accessory protein 1
There are no transcripts associated with this gene.

View Full Product Details