Product Details

SNP ID
rs13411462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:70212327 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGCACAGTTTTGGTTTTTTTTTT[A/T]AACATCTTTATATTACATGTTTTAA
Phenotype
MIM: 603518
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TIA1 PubMed Links

Gene Details

Gene
TIA1
Gene Name
TIA1 cytotoxic granule-associated RNA binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022037.2 1777 UTR 3 NP_071320.2
NM_022173.2 1777 UTR 3 NP_071505.2
XM_005264527.1 1777 UTR 3 XP_005264584.1
XM_005264528.4 1777 UTR 3 XP_005264585.1
XM_005264531.1 1777 UTR 3 XP_005264588.1
XM_011533077.1 1777 UTR 3 XP_011531379.1
XM_011533078.1 1777 UTR 3 XP_011531380.1
XM_011533081.2 1777 UTR 3 XP_011531383.1
XM_011533082.2 1777 UTR 3 XP_011531384.1
XM_017004789.1 1777 UTR 3 XP_016860278.1
XM_017004790.1 1777 UTR 3 XP_016860279.1
XM_017004791.1 1777 UTR 3 XP_016860280.1
XM_017004792.1 1777 UTR 3 XP_016860281.1
XM_017004793.1 1777 UTR 3 XP_016860282.1
XM_017004794.1 1777 UTR 3 XP_016860283.1
XM_017004795.1 1777 UTR 3 XP_016860284.1
XM_017004796.1 1777 UTR 3 XP_016860285.1
XM_017004797.1 1777 UTR 3 XP_016860286.1
XM_017004798.1 1777 UTR 3 XP_016860287.1
XM_017004799.1 1777 UTR 3 XP_016860288.1
XM_017004800.1 1777 UTR 3 XP_016860289.1

View Full Product Details