Product Details

SNP ID
rs6705704
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:32366286 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTTAAGATTTTTGGGGAGGAGAG[A/G]AAACAGATTAGTTAATGCTTTTCCA
Phenotype
MIM: 605638
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BIRC6 PubMed Links

Gene Details

Gene
BIRC6
Gene Name
baculoviral IAP repeat containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016252.3 Intron NP_057336.3
XM_005264449.4 Intron XP_005264506.2
XM_005264450.4 Intron XP_005264507.2
XM_005264451.4 Intron XP_005264508.2
XM_005264452.4 Intron XP_005264509.2
XM_005264453.4 Intron XP_005264510.2
XM_005264454.4 Intron XP_005264511.2
XM_005264455.4 Intron XP_005264512.2
XM_006712054.3 Intron XP_006712117.1
XM_006712055.3 Intron XP_006712118.1
XM_006712056.3 Intron XP_006712119.1
XM_011533003.2 Intron XP_011531305.1
XM_011533005.2 Intron XP_011531307.1
XM_017004556.1 Intron XP_016860045.1
XM_017004557.1 Intron XP_016860046.1
XM_017004558.1 Intron XP_016860047.1
XM_017004559.1 Intron XP_016860048.1
XM_017004560.1 Intron XP_016860049.1

View Full Product Details