Product Details
- SNP ID
-
rs6816145
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:163497842 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TATCTTTTGGTCGGAGGAGGGCAGA[A/G]CATTATTGCGTCTGCCACAAAAAGA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TMA16
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117172618] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TMA16
- Gene Name
- translation machinery associated 16 homolog
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018352.2 |
|
Intron |
|
|
NP_060822.2 |
View Full Product Details