Product Details

SNP ID
rs11161620
Assay Type
Functionally Tested
NCBI dbSNP Submissions
56
Location
Chr.1:75789543 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGTGAGTTCTAAAATTACACTGA[A/G]ACCTTGGAGGGTAAAATTTTTATCT
Phenotype
MIM: 602105 MIM: 179080
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MSH4 PubMed Links
Additional Information
For this assay, SNP(s) [rs112787135] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MSH4
Gene Name
mutS homolog 4
There are no transcripts associated with this gene.

Gene
RABGGTB
Gene Name
Rab geranylgeranyltransferase beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004582.3 Intron NP_004573.2
Gene
SNORD45A
Gene Name
small nucleolar RNA, C/D box 45A
There are no transcripts associated with this gene.

Gene
SNORD45B
Gene Name
small nucleolar RNA, C/D box 45B
There are no transcripts associated with this gene.

Gene
SNORD45C
Gene Name
small nucleolar RNA, C/D box 45C
There are no transcripts associated with this gene.

View Full Product Details