Product Details

SNP ID
rs12239411
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:46210071 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTGCAGTTATTTATGTTCATCTCC[C/T]CTTATTGAGACTGCAAGCTTCTATT
Phenotype
MIM: 616129 MIM: 606822
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LURAP1 PubMed Links

Gene Details

Gene
LURAP1
Gene Name
leucine rich adaptor protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013615.2 Intron NP_001013633.1
Gene
POMGNT1
Gene Name
protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243766.1 Intron NP_001230695.1
NM_001290129.1 Intron NP_001277058.1
NM_001290130.1 Intron NP_001277059.1
NM_017739.3 Intron NP_060209.3
XM_005271010.1 Intron XP_005271067.1
XM_006710755.1 Intron XP_006710818.1
XM_006710756.1 Intron XP_006710819.1
XM_011541760.2 Intron XP_011540062.1
XM_017001690.1 Intron XP_016857179.1

View Full Product Details