Product Details
- SNP ID
-
rs13265502
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:124974514 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATAATAGGCAGTAATGGTGAAGAT[A/C]CAATATAAGTACATATACTATTTTT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF572
PubMed Links
Gene Details
- Gene
- ZNF572
- Gene Name
- zinc finger protein 572
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152412.2 |
|
Intron |
|
|
NP_689625.2 |
View Full Product Details