Product Details

SNP ID
rs12171525
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:163459263 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATACACTTAACAAAGAAAAAGCGG[G/T]TCCAGATCCAAAGAGCATTGGTGAA
Phenotype
MIM: 600936
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HMMR PubMed Links

Gene Details

Gene
HMMR
Gene Name
hyaluronan mediated motility receptor
There are no transcripts associated with this gene.

Gene
NUDCD2
Gene Name
NudC domain containing 2
There are no transcripts associated with this gene.

View Full Product Details