Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_022725.3 | 699 | Intron | NP_073562.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001143830.1 | 699 | Intron | NP_001137302.1 | ||
NM_005256.3 | 699 | Intron | NP_005247.1 | ||
NM_177553.2 | 699 | Intron | NP_808221.1 | ||
XM_011519971.2 | 699 | Intron | XP_011518273.1 | ||
XM_011519972.2 | 699 | UTR 5 | XP_011518274.1 | ||
XM_011519975.1 | 699 | Intron | XP_011518277.1 | ||
XM_011519978.2 | 699 | Intron | XP_011518280.1 | ||
XM_017017528.1 | 699 | Intron | XP_016873017.1 | ||
XM_017017529.1 | 699 | Intron | XP_016873018.1 | ||
XM_017017530.1 | 699 | Intron | XP_016873019.1 | ||
XM_017017531.1 | 699 | Intron | XP_016873020.1 | ||
XM_017017532.1 | 699 | Intron | XP_016873021.1 |