Product Details

SNP ID
rs11220217
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125787198 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCAATATATTTCAATGATCATAT[A/C]ATTCCTACAACTAAAGAACATGTTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PATE2 PubMed Links

Gene Details

Gene
PATE2
Gene Name
prostate and testis expressed 2
There are no transcripts associated with this gene.

Gene
PATE3
Gene Name
prostate and testis expressed 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129883.3 1203 Intron NP_001123355.3
XM_017017054.1 1203 UTR 5 XP_016872543.1

View Full Product Details