Product Details

SNP ID
rs12721508
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:66179413 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTGAATGCAAACTCAATGAGAGG[C/G]ATCCATTCTAACTTCCTAATCTATG
Phenotype
MIM: 122560 MIM: 606469
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CRH PubMed Links

Gene Details

Gene
CRH
Gene Name
corticotropin releasing hormone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000756.3 Intron NP_000747.1
XM_017013090.1 Intron XP_016868579.1
XM_017013091.1 Intron XP_016868580.1
XM_017013092.1 Intron XP_016868581.1
XM_017013093.1 Intron XP_016868582.1
Gene
TRIM55
Gene Name
tripartite motif containing 55
There are no transcripts associated with this gene.

View Full Product Details