Product Details
- SNP ID
-
rs12112854
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:122319780 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGTCCGAACTGACTTCTCACAATG[C/T]TGCATTATGAGAAATATGTATCTCG
- Phenotype
-
MIM: 609978
MIM: 613301
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CADPS2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75643376] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CADPS2
- Gene Name
- calcium dependent secretion activator 2
- Gene
- FEZF1
- Gene Name
- FEZ family zinc finger 1
There are no transcripts associated with this gene.
- Gene
- FEZF1-AS1
- Gene Name
- FEZF1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details