Product Details

SNP ID
rs4147631
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:123548315 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACATTTCTAAGGAACCTTATGTT[C/G]CAGGTACCATGCTAGAGTCTAAGGA
Phenotype
MIM: 601677
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NDUFA5 PubMed Links

Gene Details

Gene
NDUFA5
Gene Name
NADH:ubiquinone oxidoreductase subunit A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282419.2 Intron NP_001269348.1
NM_001282420.2 Intron NP_001269349.1
NM_001282421.2 Intron NP_001269350.1
NM_001282422.2 Intron NP_001269351.1
NM_001291304.1 Intron NP_001278233.1
NM_005000.4 Intron NP_004991.1

View Full Product Details