Product Details

SNP ID
rs4731349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:127345424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAGGGAGGGTCTACTTTCAATA[C/G]AGTGCTCTGTGCAGAAATATAACAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF800 PubMed Links

Gene Details

Gene
ZNF800
Gene Name
zinc finger protein 800
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176814.4 5416 Intron NP_789784.2
XM_005250181.3 5416 Intron XP_005250238.1
XM_006715867.3 5416 UTR 3 XP_006715930.1
XM_006715868.3 5416 UTR 3 XP_006715931.1
XM_006715869.3 5416 UTR 3 XP_006715932.1
XM_011515859.2 5416 UTR 3 XP_011514161.1

View Full Product Details