Product Details

SNP ID
rs12801285
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59762821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTTGAATCTCCTTGAATGGCTC[C/G]TGGGATGAGAAAGAAAGCAGGAAGA
Phenotype
MIM: 600876
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
STX3 PubMed Links

Gene Details

Gene
STX3
Gene Name
syntaxin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178040.1 Intron NP_001171511.1
NM_004177.4 Intron NP_004168.1
XM_005274195.3 Intron XP_005274252.1
XM_005274198.2 Intron XP_005274255.1
XM_005274200.3 Intron XP_005274257.1
XM_011545221.2 Intron XP_011543523.1
XM_017018188.1 Intron XP_016873677.1
XM_017018189.1 Intron XP_016873678.1
XM_017018190.1 Intron XP_016873679.1
XM_017018191.1 Intron XP_016873680.1
XM_017018192.1 Intron XP_016873681.1
XM_017018193.1 Intron XP_016873682.1
XM_017018194.1 Intron XP_016873683.1
XM_017018195.1 Intron XP_016873684.1
XM_017018196.1 Intron XP_016873685.1

View Full Product Details