Product Details

SNP ID
rs12801213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62734880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAAAATCAGTAAATATTGAAGTA[C/G]ATTACAAGATATATGAATTTTTTGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HNRNPUL2 PubMed Links

Gene Details

Gene
HNRNPUL2
Gene Name
heterogeneous nuclear ribonucleoprotein U like 2
There are no transcripts associated with this gene.

Gene
HNRNPUL2-BSCL2
Gene Name
HNRNPUL2-BSCL2 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
TTC9C
Gene Name
tetratricopeptide repeat domain 9C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318812.1 Intron NP_001305741.1
NM_001318813.1 Intron NP_001305742.1
NM_001318814.1 Intron NP_001305743.1
NM_001318815.1 Intron NP_001305744.1
NM_001318816.1 Intron NP_001305745.1
NM_173810.3 Intron NP_776171.1

View Full Product Details