Product Details

SNP ID
rs41562314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30490437 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGCCTCTGAGGCGGAGCACCAG[A/G]GAGCCTACCTGGAAGACACATGCGT
Phenotype
MIM: 143010
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HLA-E PubMed Links

Gene Details

Gene
HLA-E
Gene Name
major histocompatibility complex, class I, E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005516.5 670 Missense Mutation AGA,GGA R178G NP_005507.3
XM_017010807.1 670 Missense Mutation AGA,GGA R219G XP_016866296.1
XM_017010808.1 670 Missense Mutation AGA,GGA R219G XP_016866297.1
XM_017010809.1 670 Missense Mutation AGA,GGA R178G XP_016866298.1

View Full Product Details