Product Details

SNP ID
rs12265962
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:60027793 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTCTCTGCCTCAGTAGAATTTAGA[G/T]TCACGCAGTGTATGGGTTGCAAATT
Phenotype
MIM: 600465
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANK3 PubMed Links

Gene Details

Gene
ANK3
Gene Name
ankyrin 3, node of Ranvier (ankyrin G)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001149.3 15633 UTR 3 NP_001140.2
NM_001204403.1 15633 UTR 3 NP_001191332.1
NM_001204404.1 15633 UTR 3 NP_001191333.1
NM_001320874.1 15633 UTR 3 NP_001307803.1
NM_020987.4 15633 UTR 3 NP_066267.2
XM_005269715.3 15633 Intron XP_005269772.1
XM_006717796.3 15633 Intron XP_006717859.1
XM_006717802.3 15633 Intron XP_006717865.1
XM_011539708.2 15633 Intron XP_011538010.1
XM_011539709.2 15633 Intron XP_011538011.1
XM_017016110.1 15633 Intron XP_016871599.1
XM_017016111.1 15633 Intron XP_016871600.1
XM_017016112.1 15633 Intron XP_016871601.1
XM_017016113.1 15633 Intron XP_016871602.1
XM_017016114.1 15633 Intron XP_016871603.1
XM_017016115.1 15633 Intron XP_016871604.1
XM_017016116.1 15633 Intron XP_016871605.1
XM_017016117.1 15633 Intron XP_016871606.1
XM_017016118.1 15633 Intron XP_016871607.1
XM_017016119.1 15633 Intron XP_016871608.1
XM_017016120.1 15633 Intron XP_016871609.1
XM_017016121.1 15633 Intron XP_016871610.1
XM_017016122.1 15633 Intron XP_016871611.1
XM_017016123.1 15633 Intron XP_016871612.1
XM_017016124.1 15633 Intron XP_016871613.1
XM_017016125.1 15633 Intron XP_016871614.1
XM_017016126.1 15633 Intron XP_016871615.1
XM_017016127.1 15633 Intron XP_016871616.1
XM_017016128.1 15633 Intron XP_016871617.1
XM_017016129.1 15633 Intron XP_016871618.1
XM_017016130.1 15633 Intron XP_016871619.1
XM_017016131.1 15633 Intron XP_016871620.1
XM_017016132.1 15633 Intron XP_016871621.1
XM_017016133.1 15633 Intron XP_016871622.1
XM_017016134.1 15633 Intron XP_016871623.1
XM_017016135.1 15633 Intron XP_016871624.1
XM_017016136.1 15633 Intron XP_016871625.1
XM_017016137.1 15633 Intron XP_016871626.1
XM_017016138.1 15633 Intron XP_016871627.1
XM_017016139.1 15633 Intron XP_016871628.1
XM_017016140.1 15633 Intron XP_016871629.1
XM_017016141.1 15633 Intron XP_016871630.1
XM_017016142.1 15633 Intron XP_016871631.1
XM_017016143.1 15633 Intron XP_016871632.1
XM_017016144.1 15633 Intron XP_016871633.1
XM_017016145.1 15633 Intron XP_016871634.1
XM_017016146.1 15633 Intron XP_016871635.1
XM_017016147.1 15633 Intron XP_016871636.1
XM_017016148.1 15633 Intron XP_016871637.1
XM_017016149.1 15633 Intron XP_016871638.1
XM_017016150.1 15633 Intron XP_016871639.1

View Full Product Details