Product Details

SNP ID
rs7079455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:60029942 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAAGCCTTACTTGCCATTCAATA[A/G]AAAAGTCTTAGGGAATAGAGGCCCT
Phenotype
MIM: 600465
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANK3 PubMed Links

Gene Details

Gene
ANK3
Gene Name
ankyrin 3, node of Ranvier (ankyrin G)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001149.3 Intron NP_001140.2
NM_001204403.1 Intron NP_001191332.1
NM_001204404.1 Intron NP_001191333.1
NM_001320874.1 Intron NP_001307803.1
NM_020987.4 Intron NP_066267.2
XM_005269715.3 Intron XP_005269772.1
XM_006717796.3 Intron XP_006717859.1
XM_006717802.3 Intron XP_006717865.1
XM_011539708.2 Intron XP_011538010.1
XM_011539709.2 Intron XP_011538011.1
XM_017016110.1 Intron XP_016871599.1
XM_017016111.1 Intron XP_016871600.1
XM_017016112.1 Intron XP_016871601.1
XM_017016113.1 Intron XP_016871602.1
XM_017016114.1 Intron XP_016871603.1
XM_017016115.1 Intron XP_016871604.1
XM_017016116.1 Intron XP_016871605.1
XM_017016117.1 Intron XP_016871606.1
XM_017016118.1 Intron XP_016871607.1
XM_017016119.1 Intron XP_016871608.1
XM_017016120.1 Intron XP_016871609.1
XM_017016121.1 Intron XP_016871610.1
XM_017016122.1 Intron XP_016871611.1
XM_017016123.1 Intron XP_016871612.1
XM_017016124.1 Intron XP_016871613.1
XM_017016125.1 Intron XP_016871614.1
XM_017016126.1 Intron XP_016871615.1
XM_017016127.1 Intron XP_016871616.1
XM_017016128.1 Intron XP_016871617.1
XM_017016129.1 Intron XP_016871618.1
XM_017016130.1 Intron XP_016871619.1
XM_017016131.1 Intron XP_016871620.1
XM_017016132.1 Intron XP_016871621.1
XM_017016133.1 Intron XP_016871622.1
XM_017016134.1 Intron XP_016871623.1
XM_017016135.1 Intron XP_016871624.1
XM_017016136.1 Intron XP_016871625.1
XM_017016137.1 Intron XP_016871626.1
XM_017016138.1 Intron XP_016871627.1
XM_017016139.1 Intron XP_016871628.1
XM_017016140.1 Intron XP_016871629.1
XM_017016141.1 Intron XP_016871630.1
XM_017016142.1 Intron XP_016871631.1
XM_017016143.1 Intron XP_016871632.1
XM_017016144.1 Intron XP_016871633.1
XM_017016145.1 Intron XP_016871634.1
XM_017016146.1 Intron XP_016871635.1
XM_017016147.1 Intron XP_016871636.1
XM_017016148.1 Intron XP_016871637.1
XM_017016149.1 Intron XP_016871638.1
XM_017016150.1 Intron XP_016871639.1

View Full Product Details