Product Details

SNP ID
rs12266039
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:75446215 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAAAAGAACCAAGGCTTTGGGGTT[A/G]GACCATCCTGGCTTGAAACCCTGCT
Phenotype
MIM: 614537
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C10orf11 PubMed Links

Gene Details

Gene
C10orf11
Gene Name
chromosome 10 open reading frame 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305581.1 Intron NP_001292510.1
NM_032024.4 Intron NP_114413.1
XM_011540256.2 Intron XP_011538558.1
XM_011540257.2 Intron XP_011538559.1
XM_017016759.1 Intron XP_016872248.1
XM_017016760.1 Intron XP_016872249.1
XM_017016761.1 Intron XP_016872250.1
XM_017016762.1 Intron XP_016872251.1

View Full Product Details