Product Details

SNP ID
rs10868132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:84275677 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTGTTTCACTGGGAGAAGGGCCAC[G/T]TACGACGTTCACAGGGTATTGGTGA
Phenotype
MIM: 608269
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC28A3 PubMed Links

Gene Details

Gene
SLC28A3
Gene Name
solute carrier family 28 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199633.1 4668 Intron NP_001186562.1
NM_022127.2 4668 Intron NP_071410.1
XM_011518905.2 4668 UTR 3 XP_011517207.1
XM_011518906.2 4668 UTR 3 XP_011517208.1
XM_011518907.2 4668 UTR 3 XP_011517209.1
XM_011518908.2 4668 UTR 3 XP_011517210.1
XM_011518909.2 4668 Intron XP_011517211.1
XM_011518910.2 4668 Intron XP_011517212.1

View Full Product Details