Product Details

SNP ID
rs10858200
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136008993 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAAGGGGCACGGGACATTGTGCGG[A/G]CCTGAACAGCAGTCAGGCGTCCTCT
Phenotype
MIM: 615786
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NACC2 PubMed Links

Gene Details

Gene
NACC2
Gene Name
NACC family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144653.4 4443 UTR 3 NP_653254.1
XM_011518221.2 4443 UTR 3 XP_011516523.1

View Full Product Details