Product Details

SNP ID
hCV31374761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:81953960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTATTTGGGAGGTGGTAGTTTA[A/G]ATAAGTTATTTGGGCTCCCTGTGGT
Phenotype
MIM: 114204
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CACNA2D1 PubMed Links

Gene Details

Gene
CACNA2D1
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 1
There are no transcripts associated with this gene.

View Full Product Details