Product Details

SNP ID
rs11666555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:22752284 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAATTTAGTTTTATATTTCAAAG[G/T]GACTAAAAGTGTAGATTTGGATTGC
Phenotype
MIM: 603981
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF99 PubMed Links

Gene Details

Gene
ZNF99
Gene Name
zinc finger protein 99
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080409.2 Intron NP_001073878.2

View Full Product Details