Product Details

SNP ID
rs13334967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:11822505 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGCTGAAAGCCTCATTACAATGCT[A/T]CCCTATCCTTCTCCATTTTTTCTTT
Phenotype
MIM: 613746
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
BCAR4 PubMed Links

Gene Details

Gene
BCAR4
Gene Name
breast cancer anti-estrogen resistance 4 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details