Product Details

SNP ID
rs13335460
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:85039056 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGGACATAAGCTCCCTGAGGGCCA[A/G]GCTTTTGTCTCTTTCACCCGGAGCA
Phenotype
MIM: 611675
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA0513 PubMed Links

Gene Details

Gene
KIAA0513
Gene Name
KIAA0513
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286565.1 Intron NP_001273494.1
NM_001286566.1 Intron NP_001273495.1
NM_001297766.1 Intron NP_001284695.1
NM_014732.3 Intron NP_055547.1
XM_005256265.3 Intron XP_005256322.1
XM_017023912.1 Intron XP_016879401.1

View Full Product Details